Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C18H15NO3
CAS Number:
Molecular Weight:
293.32
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
powder
Quality Segment
assay
≥98% (HPLC)
form
powder
color
white to beige
solubility
DMSO: 2 mg/mL, clear
storage temp.
2-8°C
Biochem/physiol Actions
PEO-IAA is an α-alkyl-indole-3-acetic acid (α-alkyl-IAA) class auxin antagonist that targets the auxin-binding site of TIR1/AFBs receptors. PEO-IAA is more potent than t-butoxycarbonylaminohexyl-IAA (BH-IAA) against auxin-dependent plant responses mediated via SCF TIR1/AFBs, including auxin-responsive gene expression, cell division and elongation pathway.
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 3 Oral
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
