Skip to Content

SML4308

Megestrol acetate

≥98% (HPLC), powder, progestogen receptor agonist

Synonym(s):

17-(Acetyloxy)-6-methylpregna-4,6-diene-3,20-dione, 6-Dehydro-6-methyl-17α-acetoxyprogesterone, 6-Methyl-17α-acetoxy-4,6-pregnadiene-3,20-dione, Megestryl acetate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C24H32O4
CAS Number:
Molecular Weight:
384.51
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

-10 to -25°C

Biochem/physiol Actions

Megestrol acetate, a well-absorbed oral synthetic progestogen, targets progesterone receptor (PR) (~130% progesterone affinity), androgen receptor (AR) (~5% metribolone affinity, antiandrogenic), and glucocorticoid receptor (GR) (~30% dexamethasone affinity). It serves as a potent progesterone receptor (PR) agonist/androgen receptor (AR) antagonist for hormone cancers and cachexia. This dual action supports its use in hormone-sensitive cancers (breast, endometrial, prostate) by inhibiting proliferation and estrogenic effects, and in prostate cancer via potential 5α reductase inhibition. Clinically, megestrol acetate acts as an antiandrogen and potent appetite stimulant, with an unclear mechanism involving Neuropeptide Y. Megestrol acetate’s efficacy in palliative cancer care and cachexia management, sometimes synergistic with tamoxifen, is tempered by potential GR-related side effects.


Still not finding the right product?

Explore all of our products under Megestrol acetate


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Repr. 1B

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library