Skip to Content

SML4310

MS-275

≥98% (HPLC), powder, HDAC inhibitor

Synonym(s):

Entinostat, MS 27-275, SNDX 275, N-[[4-[[(2-Aminophenyl)amino]carbonyl]phenyl]methyl]-carbamic acid 3-pyridinylmethyl ester, (Pyridin-3-yl)methyl 4-(2-aminophenylcarbamoyl)benzylcarbamate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C21H20N4O3
CAS Number:
Molecular Weight:
376.41
MDL number:
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

General description

MS-275 belongs to the class of 2-aminophenyl benzamides. It is a long-acting, orally bioavailable, cell-permeable, brain-region selective Class I Histone Deacetylases (HDACs) inhibitor.

Application

MS-275 is suitable for use in nano-formulation and encapsulation to study its anticancer activity.It may be used to study its therapeutic effect and regulatory mechanism in angiotensin (Ang) II-induced hypertensive mice.

Biochem/physiol Actions

MS-275 (Entinostat), a Class I Histone Deacetylases HDACs (HDAC1 IC50 ~0.18-0.51 μM; HDAC3 IC50 ~0.74-1.7 μM) inhibitor, acts via reversible active site zinc binding. This inhibition induces histone hyperacetylation, promoting euchromatin formation and DNA accessibility to transcription factors, which reactivates silenced tumor suppressor and cancer-associated genes, mediating anti-proliferative, pro-apoptotic, and differentiation-inducing effects, ultimately inhibiting cancer cell growth and survival (IC50 ~0.0415-1.92 μM in human tumor cell lines). Furthermore, MS-275 demonstrates significant in vivo antitumor activity (e.g., 5 mg/kg), exhibits immunomodulation including Treg suppression, sensitizes cancer cells to other therapies, and modulates the tumor microenvironment.


pictograms

Skull and crossbonesHealth hazard

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral - Repr. 1A

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library