Skip to Content

SML4311

Biotin Alkyne

≥95% (HPLC)

Synonym(s):

(3aS,4S,6aR)-Hexahydro-2-oxo-N-2-propyn-1-yl-1H-thieno[3,4-d]imidazole-4-pentanamide, Biotin propargylamide, N-Propargylbiotinamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C13H19N3O2S
CAS Number:
Molecular Weight:
281.37
MDL number:
Assay:
≥95% (HPLC)
Form:
powder
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

-10 to -25°C

General description

Enables selective biotinylation of azide-containing biomolecules via CuAAC for robust detection, purification, and immobilization.
Biotin Alkyne (N-Propargylbiotinamide) is a non-cleavable, alkyne-activated biotinylation reagent that enables selective labeling of azide-containing biomolecules via copper(I)-catalyzed azide-alkyne cycloaddition (CuAAC). The terminal alkyne reacts with azides to form a stable triazole linkage, facilitating permanent biotin conjugation to proteins, nucleic acids, or other targets. Biotin provides high-affinity interactions with streptavidin or avidin for detection, purification, or immobilization and, being preferentially taken up by cancer cells, serves as an excellent guiding molecule to tumor cells, enabling targeted detection and delivery. This reagent enables site-specific bioconjugation on cell surfaces, in lysates, or in vitro, excelling in bio- and chemical sensing to deliver reliable results in proteomics and molecular biology workflows.

Disclaimer

Hygroscopic Store with dessicant


Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library