Skip to Content

SML4313

Devernalizer DVR06

≥98% (HPLC)

Synonym(s):

1,3-Diazaspiro[4.4]nonan-2,4-dione, 5,5-Tetramethylenehydantoin, 5,5′-Cyclopentanespirohydantoin, Hydantoin-5-spirocyclopentane, DVR06, NSC1024, NSC 1024

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C7H10N2O2
CAS Number:
Molecular Weight:
154.17
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

General description

Non-toxic spiro-hydantoin, reverses plant vernalization epigenetically by reducing H3K27me3 at FLC, enabling heat-free control over flowering time.
DVR06, identified in Arabidopsis thaliana, induces devernalization, reversing the vernalized state at ambient temperatures. Its core function is reactivating the floral repressor FLC within the nucleus, notably in vascular and meristematic tissues. Mechanistically, DVR06 reduces repressive H3K27me3 histone methylation at the FLC locus and other genomic sites, while leaving active H3K4me3 marks unchanged. This epigenetic modulation delays flowering time and enhances vegetative growth. Possessing low toxicity and key hydantoin/spiro pharmacophores, DVR06 serves as a valuable chemical probe for epigenetic memory and holds potential for agricultural control over plant life cycles.


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Skin Sens. 1

Storage Class

11 - Combustible Solids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library