Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Assay:
≥98% (HPLC)
Form:
powder
Quality Segment
assay
≥98% (HPLC)
form
powder
color
white to beige
solubility
DMSO: 2 mg/mL, clear (warmed)
storage temp.
-10 to -25°C
Biochem/physiol Actions
1,4-DPCA potently inhibits collagen prolyl-4-hydroxylase (P4H; IC50 ~ 2 µM), HIF-prolyl hydroxylase, and TET1/TET2 enzymes (pIC50 5.42/5.26), impacting collagen biosynthesis, hypoxia signaling, and DNA demethylation. Through 2-oxoglutarate competition, it reduces proline hydroxylation, stabilizes HIF-1α protein levels, affecting collagen maturation, decreasing CD44+/CD24-/low cancer stem cells, and modulating α-KG/succinate levels. Consequently, 1,4-DPCA demonstrates significant anti-cancer effects in breast cancer models, inhibiting tumor growth, metastasis, and altering cellular metabolism via downregulation of HIF-1 targets LDHA/PDK1. FG-0041 in combination with Y-27632 initiates differentiation of mesenchymal stem cells into neuron-like cells. Notably, DPCA enhances CAR-T cell expansion and function, improving chemotherapeutic efficacy, potentially by affecting oxygen consumption, extracellular acidification, and reactive oxygen species levels.
signalword
Warning
hcodes
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2
Storage Class
11 - Combustible Solids
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
