Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Assay:
≥98% (HPLC)
Form:
powder
Quality Segment
assay
≥98% (HPLC)
form
powder
color
white to beige
solubility
DMSO: 2 mg/mL, clear
storage temp.
-10 to -25°C
Biochem/physiol Actions
Enobosarm (Ostarine) is a potent androgen receptor (AR) partial agonist (Ki = 3.8 nM) that exhibits anabolic activity in the body (bone and muscle mass), with minimal effects on the gonadal tissues. Enobosarm is a SARM (selective androgen receptor modulator) in clinical development for prevention and treatment of muscle wasting in patients with cancer.
signalword
Warning
hcodes
Hazard Classifications
Repr. 2
Storage Class
11 - Combustible Solids
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
