Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Assay:
≥98% (HPLC)
Form:
powder
Quality Segment
assay
≥98% (HPLC)
form
powder
color
white to beige
solubility
DMSO: 2 mg/mL
storage temp.
-10 to -25°C
Biochem/physiol Actions
Esaxerenone (CS-3150) is a selective, reversible, non-steroidal, orally available, cell-permeable mineralocorticoid receptor (MR) antagonist, functioning as a "silent antagonist." Selectively binds the MR ligand-binding domain with high affinity (IC50 human ~3.7 nM for blocking aldosterone-induced transcription), inhibiting MR activation and downstream gene transcription, including profibrotic/pro-inflammatory markers. Esaxerenone exhibits over 1,000-fold selectivity for MR over other NR3C receptors, minimizing endocrine side effects compared to steroidal MRAs. Its core actions involve dose-dependent sodium/water reduction and increased potassium excretion, leading to lower blood pressure and albuminuria. Consequently, esaxerenone effectively treats hypertension and diabetic nephropathy, offering cardio-renal protection with an improved safety profile.
signalword
Danger
hcodes
Hazard Classifications
Eye Dam. 1 - Skin Sens. 1B
Storage Class
11 - Combustible Solids
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

