Skip to Content

SML4338

Nicotinic acid riboside

≥95% (HPLC)

Synonym(s):

3-Carboxy-1-β-D-ribofuranosyl-pyridinium hydroxide inner salt, Nicotinic riboside, Nicotinate D-ribonucleoside, Nicotinic acid ribonucleoside

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C11H13NO6
CAS Number:
Molecular Weight:
255.22
Assay:
≥95% (HPLC)
Form:
powder
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

Nicotinic acid riboside (NAR, NR), an orally bioavailable Vitamin B3 derivative and authentic nicotinamide adenine dinucleotide (NAD+) intermediate, primarily functions to elevate intracellular NAD+ levels. Following cellular uptake via transporters, NAR is converted to NAD+ through the Preiss-Handler pathway, initiated by NR kinases. This NAD+ increase is crucial as NAD+ is vital for essential cellular processes like energy metabolism, DNA repair, and sirtuin activation, which are often impaired with age. Consequently, NAR holds significant therapeutic potential by restoring NAD+ to support cellular health and mitigate age-related and metabolic dysfunction, thereby enhancing cellular resilience and function.

Disclaimer

Hygroscopic Store with desiccant


Storage Class

11 - Combustible Solids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library