Skip to Content

SML4364

AMXT1501 tetrahydrochloride

≥98% (HPLC)

Synonym(s):

(R)-N-(5-amino-6-((3-((4-((3-aminopropyl)amino)butyl)amino)propyl)amino)-6-oxohexyl)palmitamide tetrahydrochloride, D-Lys(C16acyl)-Spm 4HC, AMXT 1501 4HC, AMXT-1501 4HCl

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C32H68N6O2·4HCl
CAS Number:
Molecular Weight:
714.77
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 2 mg/mL, clear (warmed)

storage temp.

-10 to -25°C

Biochem/physiol Actions

AMXT1501 is a lipophilic polyamine analog that acts as a potent polyamine transport inhibitor. When combined with the polyamine biosynthesis inhibitor DFMO, AMXT1501 potently inhibits cell growth in cancer cultures (GI50 in nM in the presence of 5 mM DFMO, 500 nM spermidine & 1 mM aminoguanidine = 76/MDA-231, 120/PC-3, 31/A375, 170/SK-OV-3) and induces complete or nearly complete remission of cutaneous squamous cell carcinomas (SCC) growth in a transgenic ornithine decarboxylase (ODC) mouse model of skin cancer in vivo (0.5-0.7 mg/kg via b.i.d. i.p. with DFMO at 0.5% in drinking water).

Disclaimer

Hygroscopic, store with desiccant


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library