Skip to Content

68415

2-(Nitrosomethylamino)pyridine

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

N-Methyl-N-nitroso-2-pyridinamine, N-Nitroso-N-methyl-2-aminopyridine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C6H7N3O
CAS Number:
Molecular Weight:
137.14
UNSPSC Code:
41116107
NACRES:
NA.21
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

grade

certified reference material, TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

format

neat

storage temp.

−20°C

SMILES string

N(N=O)(C)c1ncccc1

InChI

1S/C6H7N3O/c1-9(8-10)6-4-2-3-5-7-6/h2-5H,1H3

InChI key

WJDNMDMNPIKDCP-UHFFFAOYSA-N

General description

Nitrosamines are chemical compounds that can form during drug manufacturing and may pose health risks, including carcinogenicity.

Application

2-(Nitrosomethylamino)pyridine solution is commonly employed as a reference compound in analytical chemistry. It is utilized in chromatographic and spectrometric techniques for the detection and quantification of nitrosamines in chemical, pharmaceutical, and environmental matrices.

Analysis Note

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with  ISO 17034  and  ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under 2-(Nitrosomethylamino)pyridine


pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Carc. 2 - Muta. 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library