Skip to Content

73992

2-Aminofluorene

VETRANAL®, analytical standard

Synonym(s):

2-Fluorenamine, 2-Fluorenylamine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C13H11N
CAS Number:
Molecular Weight:
181.23
UNSPSC Code:
41116107
Beilstein/REAXYS Number:
1945861
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

product line

VETRANAL®

mp

124-128 °C (lit.)

application(s)

food and beverages
pharmaceutical

format

neat

SMILES string

Nc1ccc-2c(Cc3ccccc-23)c1

InChI

1S/C13H11N/c14-11-5-6-13-10(8-11)7-9-3-1-2-4-12(9)13/h1-6,8H,7,14H2

InChI key

CFRFHWQYWJMEJN-UHFFFAOYSA-N

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under 2-Aminofluorene


Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Codes

Hazard Classifications

Aquatic Chronic 2 - Carc. 2

Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library