Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C19H30O3
CAS Number:
Molecular Weight:
306.44
MDL number:
NACRES:
NA.21
grade
analytical standard
Quality Segment
assay
≥98.0% (HPLC)
technique(s)
HPLC: suitable, gas chromatography (GC): suitable
application(s)
cosmetics
food and beverages
format
neat
storage temp.
2-8°C
SMILES string
O(CC)C(=O)CCc1cc(c(c(c1)C(C)(C)C)O)C(C)(C)C
InChI
1S/C19H30O3/c1-8-22-16(20)10-9-13-11-14(18(2,3)4)17(21)15(12-13)19(5,6)7/h11-12,21H,8-10H2,1-7H3
InChI key
ZCWSUZJGZZFSHM-UHFFFAOYSA-N
Hazard Codes
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2
Storage Class
11 - Combustible Solids
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
