Skip to Content

77097

Tetramethylthiuram disulphide

analytical standard

Synonym(s):

Tetramethylthiuram disulfide, TMTD, Bis(dimethylthiocarbamoyl) disulfide, Bis(dimethylthiocarbamyl) disulfide, Thiram

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
(CH3)2NCSS2CSN(CH3)2
CAS Number:
Molecular Weight:
240.43
Beilstein/REAXYS Number:
1725821
MDL number:
UNSPSC Code:
41116107
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

product line

Certipur®

autoignition temp.

316 °F

technique(s)

HPLC: suitable, LC/MS: suitable, liquid chromatography (LC): suitable, mass spectrometry (MS): suitable

mp

156-158 °C (lit.)

application(s)

food and beverages
pharmaceutical

format

neat

SMILES string

CN(C)C(=S)SSC(=S)N(C)C

InChI

1S/C6H12N2S4/c1-7(2)5(9)11-12-6(10)8(3)4/h1-4H3

InChI key

KUAZQDVKQLNFPE-UHFFFAOYSA-N

Legal Information

CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany


Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 Oral

Target Organs

Liver

Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library