Skip to Content

93073

Isatin

Synonym(s):

2,3-Indolinedione, Isatine, NSC 9262

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C8H5NO2
CAS Number:
Molecular Weight:
147.13
Beilstein/REAXYS Number:
383659
MDL number:
NACRES:
NA.21
UNSPSC Code:
12352100
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

shelf life

limited shelf life, expiry date on the label

mp

193-195 °C (dec.) (lit.)

application(s)

food and beverages
personal care
pharmaceutical

format

neat

SMILES string

O=C1Nc2ccccc2C1=O

InChI

1S/C8H5NO2/c10-7-5-3-1-2-4-6(5)9-8(7)11/h1-4H,(H,9,10,11)

InChI key

JXDYKVIHCLTXOP-UHFFFAOYSA-N



Still not finding the right product?

Explore all of our products under Isatin


Storage Class

11 - Combustible Solids

wgk

WGK 1



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library