Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C6H9N3O2
CAS Number:
Molecular Weight:
155.15
MDL number:
NACRES:
NA.21
grade
analytical standard
Quality Segment
assay
≥98.0% (HPLC)
mp
295 °C (dec.) (lit.)
application(s)
food and beverages
pharmaceutical
plastics
format
neat
storage temp.
2-8°C
SMILES string
CN1C(N)=CC(=O)N(C)C1=O
InChI
1S/C6H9N3O2/c1-8-4(7)3-5(10)9(2)6(8)11/h3H,7H2,1-2H3
InChI key
VFGRNTYELNYSKJ-UHFFFAOYSA-N
Still not finding the right product?
Explore all of our products under 6-Amino-1,3-dimethyluracil
Storage Class
11 - Combustible Solids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.