Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C17H27NO4S
CAS Number:
Molecular Weight:
341.47
UNSPSC Code:
12352200
MDL number:
NACRES:
NA.24
grade
analytical standard
Quality Segment
product line
PESTANAL®
assay
≥90.0% (HPLC)
shelf life
limited shelf life, expiry date on the label
format
neat
storage temp.
2-8°C
Application
Metabolite for water testing
Other Notes
For R&D, and Analytical use only. Not for drug, household or other uses
Legal Information
PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany
Signal Word
Warning
Hazard Codes
Precautionary Statements
Hazard Classifications
Repr. 2
Storage Class
11 - Combustible Solids
WGK
WGK 3
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
