Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C6H6N2O3
CAS Number:
Molecular Weight:
154.12
NACRES:
NA.24
UNSPSC Code:
41116107
grade
USP Analytical Reference Materials (ARMs)
agency
USP
manufacturer/tradename
USP
mp
177-180 °C
application(s)
pharmaceutical small molecule
format
neat
storage temp.
2-8°C
InChI
1S/C6H6N2O3/c1-4-2-7-3-5(6(9)10)8(4)11/h2-3H,1H3,(H,9,10)
InChI key
DNRXJHATQULEHC-UHFFFAOYSA-N
General description
Find answers to frequently asked questions on USP products in USP FAQs.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets, have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.
Biochem/physiol Actions
Niacin-derived, vasodilator studied for its lipid-lowering effect.
Analysis Note
These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.
Still not finding the right product?
Explore all of our products under Acipimox
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.