Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C27H50O6
CAS Number:
Molecular Weight:
470.68
UNSPSC Code:
41116107
grade
pharmaceutical primary standard
agency
USP
form
liquid
manufacturer/tradename
USP
packaging
pkg of (3 x 0.5 mL)
application(s)
pharmaceutical small molecule
format
neat
storage temp.
2-30°C
SMILES string
O=C(OCC(OC(=O)CCCCCCC)COC(=O)CCCCCCC)CCCCCCC
InChI
InChI=1S/C27H50O6/c1-4-7-10-13-16-19-25(28)31-22-24(33-27(30)21-18-15-12-9-6-3)23-32-26(29)20-17-14-11-8-5-2/h24H,4-23H2,1-3H3
InChI key
VLPFTAMPNXLGLX-UHFFFAOYSA-N
General description
Find answers to frequently asked questions on USP products in USP FAQs.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets, have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.
Analysis Note
These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.
Storage Class
10 - Combustible liquids
flash_point_f
392.4 °F
flash_point_c
200.2 °C
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.