Skip to Content

1546400

USP

Polyethylene Glycol 30 Dipolyhydroxystearate 5

United States Pharmacopeia (USP) Reference Standard

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C20H40O4
CAS Number:
Molecular Weight:
344.53
UNSPSC Code:
41116107
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

form

waxy solid

manufacturer/tradename

USP

packaging

pkg of (200 mg)

application(s)

pharmaceutical small molecule

format

neat

storage temp.

2-30°C

InChI

InChI=1S/C18H36O3.C2H4O/c1-2-3-4-11-14-17(19)15-12-9-7-5-6-8-10-13-16-18(20)21;1-2-3-1/h17,19H,2-16H2,1H3,(H,20,21);1-2H2

InChI key

WOGBVVJPYOUGFE-UHFFFAOYSA-N

General description

Find answers to frequently asked questions on USP products in USP FAQs.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets, have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  

Other Notes

Sales restrictions may apply.
USP issued SDS can be found here


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library