Skip to Content
MilliporeSigma

1576720

USP

Propylene glycol diacetate

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Propylene Glycol Diacetate, 1,2-Diacetoxypropane, 1,2-Propanediol diacetate, Propylene diacetate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
CH3CH(OCOCH3)CH2OCOCH3
CAS Number:
Molecular Weight:
160.17
Beilstein/REAXYS Number:
1768914
MDL number:
NACRES:
NA.24
UNSPSC Code:
41116107
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

autoignition temp.

808 °F

manufacturer/tradename

USP

refractive index

n20/D 1.414 (lit.)

bp

191 °C (lit.)

density

1.05 g/mL at 25 °C (lit.)

application(s)

pharmaceutical small molecule

format

neat

storage temp.

2-8°C

SMILES string

CC(COC(C)=O)OC(C)=O

InChI

1S/C7H12O4/c1-5(11-7(3)9)4-10-6(2)8/h5H,4H2,1-3H3

InChI key

MLHOXUWWKVQEJB-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets, have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.

Other Notes

Sales restrictions may apply.
USP issued SDS can be found here


Still not finding the right product?

Explore all of our products under Propylene glycol diacetate




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library