Select a Size
About This Item
grade
pharmaceutical primary standard
agency
USP
form
powder
packaging
pkg of 10 mg
manufacturer/tradename
USP
application(s)
pharmaceutical small molecule
format
neat
storage temp.
−20°C
SMILES string
O=CC1OC(=O)C2=CC(O)=C(O)C(O)=C2C3=C(O)C(O)=C(O)C=C3C(=O)OC1C4OC(=O)C5=CC(O)=C(O)C(O)=C5C6=C(O)C(O)=C7OC(=O)C=8C(=C(O)C(O)=C9OC(=O)C6=C7C98)C%10=C(O)C(O)=C(O)C=C%10C(=O)OCC4O
InChI
InChI=1S/C48H28O30/c49-5-16-40(76-46(70)9-3-13(52)28(56)32(60)18(9)17-8(44(68)74-16)2-12(51)27(55)31(17)59)39-15(54)6-73-43(67)7-1-11(50)29(57)33(61)19(7)21-25-23-24-26(48(72)78-41(23)37(65)35(21)63)22(36(64)38(66)42(24)77-47(25)71)20-10(45(69)75-39)4-14(53)30(58)34(20)62/h1-5,15-16,39-40,50-66H,6H2
InChI key
SKNLUADAGHCXKF-UHFFFAOYSA-N
General description
Analysis Note
Still not finding the right product?
Explore all of our products under Punicalagin
Storage Class
11 - Combustible Solids
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.