Skip to Content

1605012

USP

Roflumilast

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

3-(Cyclopropylmethoxy)-N-(3,5-dichloro-4-pyridinyl)-4-(difluoromethoxy)benzamide, 3-Cyclopropylmethoxy-4-difluoromethoxy-N-[3,5-dichloropyrid-4-yl]-benzamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C17H14Cl2F2N2O3
CAS Number:
Molecular Weight:
403.21
MDL number:
NACRES:
NA.24
UNSPSC Code:
41116107
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

manufacturer/tradename

USP

application(s)

pharmaceutical small molecule

format

neat

storage temp.

2-8°C

SMILES string

ClC1=CN=CC(Cl)=C1NC(C2=CC=C(OC(F)F)C(OCC3CC3)=C2)=O

InChI

1S/C17H14Cl2F2N2O3/c18-11-6-22-7-12(19)15(11)23-16(24)10-3-4-13(26-17(20)21)14(5-10)25-8-9-1-2-9/h3-7,9,17H,1-2,8H2,(H,22,23,24)

InChI key

MNDBXUUTURYVHR-UHFFFAOYSA-N

General description

Find answers to frequently asked questions on USP products in USP FAQs.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets, have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Biochem/physiol Actions

Roflumilast is a highly potent, orally active, and selective phosphodiesterase 4 (PDE4) inhibitor with an IC50 of 0.8 nM. Roflumilast has anti-inflammatory properties and is used clinically to treat COPD.
Roflumilast is a highly potent, orally active, and selective phosphodiesterase 4 (PDE4) inhibitor.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.

Other Notes

Sales restrictions may apply.
USP issued SDS can be found here


Still not finding the right product?

Explore all of our products under Roflumilast


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

No data available

flash_point_c

No data available



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library