Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Linear Formula:
CH3(CH2)26CH3
CAS Number:
Molecular Weight:
394.76
Beilstein/REAXYS Number:
1770570
MDL number:
UNSPSC Code:
41116107
grade
USP Analytical Reference Materials (ARMs)
agency
USP
vapor density
13.6 (vs air)
vapor pressure
<1 mmHg ( 20 °C)
manufacturer/tradename
USP
bp
278 °C/15 mmHg (lit.)
mp
57-64 °C (lit.)
application(s)
pharmaceutical small molecule
format
neat
storage temp.
2-8°C
SMILES string
CCCCCCCCCCCCCCCCCCCCCCCCCCCC
InChI
1S/C28H58/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h3-28H2,1-2H3
InChI key
ZYURHZPYMFLWSH-UHFFFAOYSA-N
General description
Find answers to frequently asked questions on USP products in USP FAQs.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets, have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.
Analysis Note
These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.
Still not finding the right product?
Explore all of our products under Octacosane
Storage Class
11 - Combustible Solids
flash_point_f
440.6 °F
flash_point_c
227 °C
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.