Skip to Content

1A05570

USP

Atracurium Besylate Dimethylisoquinolinium Analog

Pharmaceutical Analytical Impurity (PAI)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C28H36NO7S
CAS Number:
Molecular Weight:
530.65
UNSPSC Code:
41116107
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical analytical impurity (PAI)

agency

USP

form

powder

manufacturer/tradename

USP

application(s)

pharmaceutical small molecule

format

neat

storage temp.

−20°C

SMILES string

O=S(=O)([O-])C=1C=CC=CC1.O(C1=CC=C(C=C1OC)CC2C3=CC(OC)=C(OC)C=C3CC[N+]2(C)C)C

InChI

InChI=1S/C22H30NO4.C6H6O3S/c1-23(2)10-9-16-13-21(26-5)22(27-6)14-17(16)18(23)11-15-7-8-19(24-3)20(12-15)25-4;7-10(8,9)6-4-2-1-3-5-6/h7-8,12-14,18H,9-11H2,1-6H3;1-5H,(H,7,8,9)/q+1;/p-1

InChI key

KSXQDTVGIGIGBP-UHFFFAOYSA-M

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets, have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.
Find answers to frequently asked questions on USP products in USP FAQs.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.

Other Notes

Sales restrictions may apply.
USP issued SDS can be found here.


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Hazard Classifications

Acute Tox. 4 Oral - Skin Irrit. 2

Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library