Select a Size
About This Item
grade
pharmaceutical analytical impurity (PAI)
agency
USP
form
powder
packaging
pkg of 25 mg
manufacturer/tradename
USP
application(s)
pharmaceutical small molecule
format
neat
storage temp.
-10 to -25°C
SMILES string
O=C(OC(C)(C)C)NC(C=1C=CC=CC1)C(O)C(=O)OC2C(=C3C(=O)C(=O)C4(C)C(O)CC5OCC5(OC(=O)C)C4C(OC(=O)C=6C=CC=CC6)C(O)(C2)C3(C)C)C
InChI
InChI=1S/C43H51NO14/c1-22-26(55-37(51)32(48)30(24-15-11-9-12-16-24)44-38(52)58-39(3,4)5)20-43(53)35(56-36(50)25-17-13-10-14-18-25)33-41(8,34(49)31(47)29(22)40(43,6)7)27(46)19-28-42(33,21-54-28)57-23(2)45/h9-18,26-28,30,32-33,35,46,48,53H,19-21H2,1-8H3,(H,44,52)/t26-,27-,28+,30-,32+,33-,35-,41+,42-,43+/m0/s1
InChI key
ZOLQDWANVNOXBK-YYCGEXIESA-N
General description
Analysis Note
Signal Word
Danger
Hazard Codes
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Lact. - Muta. 2 - Repr. 1B - Skin Irrit. 2 - STOT RE 1
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

