Parteck® Functional Excipients
A high-quality portfolio of Parteck® excipients for solid dosage forms
Tableting Formulation
As the solid formulation design requires a fine balance, we have developed our Parteck® excipients to beat formulation challenges with unique particle properties. These particle-engineered excipients provide individual functionalities for excellent performance in tableting processes, for specific drug delivery technologies, or for solubility enhancement. Selecting the right excipients is key for a successful formulation and with our Parteck® portfolio, you’ll experience that your solid dosage forms are easier to design. Furthermore, you will be supported by our stringent quality control and a full regulatory support.
Benefits of our Parteck® portfolio:
Flexible tableting:
Optimized drug delivery:
  • Modification of the release kinetics of your formulation
  • Excipients for different routes of administration
Enhanced solubility:
  • Boost the efficacy of your final drug product by enhancing API solubility
EMPROVE® documentation to expedite your approval preparation
Parteck® M Directly compressible mannitol, combining stability and rapid disintegration Flyer
100494 Parteck® M 100 (Mannitol) EMPROVE® ESSENTIAL Ph Eur,BP,JP,USP,E 421  
100419 Parteck® M 200 (Mannitol) EMPROVE® ESSENTIAL Ph Eur,BP,JP,USP,E 421  
Parteck® Delta M Convertible mannitol specifically designed for wet granulation Flyer
112635 Parteck® Delta M (Mannitol) EMPROVE® ESSENTIAL Ph Eur,BP,USP,JP,E 421  
Parteck® LUB A range of vegetable-origin stearates for a reliable lubrication Flyer
100661 Parteck® LUB STA 50 (Stearic acid 50 vegetable grade) EMPROVE® ESSENTIAL Ph Eur,BP,JP,NF  
100663 Parteck® LUB MST (MAGNESIUM STEARATE VEGETABLE GRADE) EMPROVE® ESSENTIAL Ph Eur,BP,JP,NF,FCC  
100664 Parteck® LUB CST (Calcium stearate vegetable grade) EMPROVE® ESSENTIAL Ph Eur,BP,JP,NF,FCC  
Parteck® SI Directly compressible sorbitol, combining excellent flowability and a great mouth feel  Flyer
115079 Parteck® SI 200 (Sorbitol) EMPROVE® ESSENTIAL Ph Eur,BP,JP,NF,E 420  
103140 Parteck® SI 400 (Sorbitol) EMPROVE® ESSENTIAL Ph Eur,BP,JSFA,NF,E 420  
103583 Parteck® SI 150 (Sorbitol) EMPROVE® ESSENTIAL Ph Eur,BP,JP,NF,JSFA,E 420  
103557 Parteck® SI 450 (Sorbitol) EMPROVE® ESSENTIAL NF,FCC,JSFA  
Parteck® CCS Superdisintegrant for solid formulations  
102310 Parteck® CCS Croscarmellose Sodium EMPROVE® ESSENTIAL Ph.Eur.,JP,NF  
Parteck® COAT Polyvinyl alcohol for rapid preparation times and full flexibility in film coatings Flyer
141517 Parteck® COAT (Polyvinyl alcohol) EMPROVE® ESSENTIAL Ph Eur,ChP,JPE,USP  
Parteck® SRP 80 Polyvinyl alcohol-based excipient for consistent, sustained drug release over long release periods Flyer
141439 Parteck® SRP 80 (Polyvinyl alcohol) EMPROVE® ESSENTIAL Ph Eur,ChP,USP,JPE  
Parteck® ODT Mannitol-based excipient system for orally disintegrating tablets Flyer
100490 Parteck® ODT EMPROVE® ESSENTIAL  
Parteck® M DPI Mannitol-based versatile alternative carrier for dry powder inhalation applications Flyer
103668 Parteck® M DPI EMPROVE® EXPERT Ph Eur,BP,JP,USP,E 421  
Parteck® SLC Silica drug carrier with a unique surface structure Flyer
120091 Parteck® SLC 500  
Parteck® MXP Polyvinyl alcohol for hot-melt extrusion, allowing for stable and high drug loads Flyer
141464 Parteck® MXP (Polyvinyl alcohol) EMPROVEParteck® ESSENTIAL Ph Eur,JPE,USP