Bulk Powder Transfer Bags
EZ BioPac™ and Right Sized Weighing for safer, faster processing.
EZ BioPAC™, from ILC Dover, for safer, faster use of cell culture media and other bioprocess chemicals. Easier to dispense to reduce potential operator injury the bags connects directly to hydration tank minimizing product contamination risk while safeguarding employees and facilities. Our bulk powder transfer bags are scalable from 1 Kg to 100 Kg and are pre-weighed to within +/-0.5% accuracy to eliminate potential for error and save time. Antistatic film properties allow for 99.9% product recovery.
Industry Leading Transfer Bag Design
EZ BioPac™ bags are easier to dispense, Reducing the potential for injury. Larger sizes include lifting straps.
Dust-free Transfer Process
Stretch or flange for a quick, secure
connection.
Complete Discharge, No Rinsing
Bag connects directly to hydration tank, minimizing product contamination risk, while safeguarding employees and facilities.
Provides Accuracy & Confidence in Weigh
Pre-weighted to within +/- 0.5% accuracy to eliminate potential for error and save time.
No Need to Reweigh
Manual weighing processes increase potential operator risk for injury and exposure to dust.
Recovery
Armorflex® 114 film construction with permanent antistatic properties for 99.9% product recovery.
Accessories and Bags
Product No. Accessory Description Quantity Add to Cart
HW11200-02 Lifting Bars Low profile lifting bar for products DP4603, DP4604
1
HW 11089 Lifting Bars Low profile lifting bar for products DP4487, DP4600, DP4537,
DP4601, DP4652
1
HW10977-01 Lifting System Stainless steel manual lifting system ≤45 Kg capacity 1
HW10506-01 Lifting System Stainless steel manual lifting system ≤50 Kg capacity 1
HW11230-01 Hand tool Stainless steel cable tie removal tool 1
DP4714-1EA Dry Bags EZ BIOPAC, 40L, v-fold neck, 3in flange, AF114 1
DP4715-1EA Dry Bags EZ BIOPAC, 40L, v-fold neck, 3in non gasket, AF114 1
Download Data Sheet
View the technical data sheet for the EZ BioPac™ bulk powder transfer
bags
Manufacturer Drawing
View the manufacturer drawing for the EZ BioPac bulk powder transfer bags
View Comparison Flowchart
Compare the advantages of using the EZ BioPac bulk powder transfer bags from ILC Dover over the standard process.
Want More Information?
   Contact Us