ESHMUNO® CP-FT Resin
Explore Efficiency
The first strong cation exchange chromatography resin that is specifically designed for efficient mAb aggregate removal in flow-through mode.
Benefits
Increased performance
  • Superior removal of mAb aggregates in the flow-through
    mode of operation
  • High product recoveries at high mass loadings
Reduced costs
  • Significant reduction in resin and buffer volume
  • Smaller manufacturing footprint (smaller columns, buffer
    tanks, etc.)
Intensified process
  • Low salt process conditions eliminate the need for dilution
    before subsequent ion exchange steps
  • Processing volume reduction for later virus filtration and
    ultrafiltration steps
Enhanced ease of use
  • Rigid base beads enable higher flow rates and
    easier column packing
Eshmuno® CP-FT Resin
Novel tentacular ligands developed
Figure 1: Novel Tentacular ligands developed
ESHMUNO® CP-FT Resin
Product No. Description
120093 ESHMUNO® CP-FT
125168 MiniChrom Column ESHMUNO® CP-FT, 1 ml 8x20mm
125169 MiniChrom Column ESHMUNO® CP-FT, 5 ml 8x100mm
125170 MiniChrom Column ESHMUNO® CP-FT, 0.2 ml 5x10mm
125171 RoboColumn ESHMUNO® CP-FT, 0.2 ml 8 PC 5x10mm
125172 RoboColumn ESHMUNO® CP-FT, 0.6 ml 8 PC 5x30mm
We offer safe, integrated and innovative solutions for each of your chromatography steps from process development to production scale. See our full Chromatography portfolio, available for purchase:
Affinity Chromatography
Product No. Description
120089 ESHMUNO® A
120094 ESHMUNO® P anti-A
120095 ESHMUNO® P anti-B
Ion Exchange Chromatography
Product No. Description
116887 Fractogel® EMD TMAE (S)
116884 Fractogel® EMD DMAE (M)
114894 Fractogel® EMD SE Hicap (M)
116885 Fractogel® EMD TMAE Medcap (M)
116886 Fractogel® EMD COO⁻ (M)
110316 Fractogel® EMD TMAE Hicap (M)
110338 Fractogel® EMD Chelate (M)
110317 Fractogel® EMD BioSEC (S)
116890 Fractogel® EMD SO₃⁻ (S)
116882 Fractogel® EMD SO₃⁻ (M)
116883 Fractogel® EMD DEAE (M)
116881 Fractogel® EMD TMAE (M)
120087 ESHMUNO® HCX
120079 ESHMUNO® Q
120083 ESHMUNO® CPX
120078 ESHMUNO® S
120093 ESHMUNO® CP-FT
Membrane Chromatography
Product No. Description
NXF-01 Natrix® Q Recon Mini
NXF-10 Natrix® Q Pilot
NXF-20 Natrix® Q Process 150
NXF-50 Natrix® Q Process 600