Pellicon® Capsule
The first-of-its-kind, a single-use tangential flow filter that comes ready to process mAbs and ADCs in minutes
We have engineered the first-of-its-kind Pellicon® Capsule, a true single-use tangential flow filter that offers comparable performance to the industry leading Pellicon® cassette.
Through our innovative spiral technology, Pellicon® Capsule provides the efficiency you know and expect with the convenience of a pre-sterilized and self-contained format to easily facilitate single-use processing.
The Pellicon® Capsule simplifies ultrafiltration and diafiltration processes with its user-friendly features for improved process economics and speed to market.
Our new Pellicon® Capsule was designed to address customer needs including:
  • Plug and play, holderless format
  • Gamma sterilized and preservative-free device
  • Self-containment for product and operator safety
  • Fast and flexible product changeover