St. Louis, MO, Facility Overview
ADCs, Bio-conjugation, and Bio-organics
Our St. Louis (USA) manufacturing site has more than 30 years of experience in bio-conjugation, APIs, excipients and adjuvants manufacturing. Extensive analytical capabilities and dedicated compliance resources, along with innovative manufacturing capabilities help our customers around the globe to accelerate their drug development programs.
Bio-conjugates
(Potent bio-conjugates (ADC), non-potent bio-conjugates)
Antibody-drug conjugation technology uses monoclonal antibodies or other biologics to deliver highly potent active pharmaceutical ingredients (HPAPIs) to targeted cells. In conjugated form, the HPAPIs exhibits more selective cytotoxicity, thereby, sparing non-target cells from many of the toxic effects and improving the safety profile.
Bio-conjugates also use traditional APIs and possible new novel drugs to provide a similar therapeutic effect.
ADC Capabilities
We have the expertise needed to deliver solutions for your bioconjugation, whether it be potent (ADCs) or non-potent for clinical and commercial supplies.
  • 45 constructs and >400 development batches and >115 cGMP batches, with extensive chromatography experience
    • random cysteine or lysine conjugation technology
    • site directed conjugation via engineered mAbs or enzyme catalyzed
    • various payloads (Microtubule Inhibitors or DNA binding)
  • Analytical capabilities for characterization, including mass spectrometry and cell-bases assays
  • Release testing and stability for both Bulk Drug Substance (BDS) and Drug Product (DP)
  • Personnel and suites dedicated to ADC development, manufacturing, and testing
Potent-Bioconjugates (ADCs)
QTY Equipment Capacity Temp Range
1
Clinical ADC Suite 10-100 L 2 to 37 °C
1 Commercial ADC suite 600 L max reactor 2 to 37 °C
2 Mobius® FlexReady TFF systems up to 200 L  
4 Mobius® Single Use Mixing System 50 L to 100 L  
  WFI supply system    
1 AKTA™ Ready Chromatography System    
  Bulk Fill Room (Grade C)    
  Isolator (Grade A) with VHP for bottle filling    
  GE 6610 Autoclave    
2 Biosafety Cabinet    
Non-Potent-Bioconjugates
QTY Equipment Capacity Temp Range
2
Jacketed Reactors 2,5000 L 2 to 37 °C
2 Portable Equipment up to 1,000 L  
2 Mobius® FlexReady TFF systems up to 200 L  
4 Mobius® Single Use Mixing System 50 to 100 L  
  WFI supply system    
  AKTA™ Ready Chromatography System    
  Biosafety Cabinet    
Complex API Lab and Equipment
This facility is focused on the synthesis and purification of bio-organic materials such as polyamino acids, liposomes, polynucleotides, and lipids.
Bio-organics
QTY Equipment Capacity Temp Range
6
Glass lined reactors 50 L - 3000 Gallons -9 C to 120 °C
2 Filter Dryer 0.6 m2 to 2 m2 -9 C to 120 °C
3 Walk in Hoods 50 L reactors Plant operating temperature 17 °C
  Chromatography    
  Hastelloy Centrifuge Capacity is 120 kg Ambient
  Lyophilization 200 L -50 to +30 °C
Process and Analytical Development
Our supporting services include developing robust analytical methodology platforms supporting all cGMP manufacturing areas:
  • State-of-the-art analytical methods for characterization of bio-conjugates and complex APIs
  • Bio-analytical methods for product functional characterization – binding, enzyme and cell based assays
Quality Management and Compliance
Our offer includes extensive regulatory expertise in quality, compliance and regulatory:
  • ICH Q7 is our Quality System and the Global standard for the manufacturing, testing, packaging, and release of APIs
  • ISO 9000 cGMP compliant operations
  • 21 CFR 210/211 is in place for contract Drug Product testing for our customers
  • FDA registered site with commercial API production since 1999
Ability to support customer development activities:
  • Preparation of regulatory filings (CMC sections)
  • Vendor audits
  • Control documentation and testing
Project Management
From evaluation to execution, our dedicated project managers are coordinating multi-disciplinary teams, international site activities and timelines throughout the lifecycle of your program.