[Skip to Content](https://www.sigmaaldrich.com#main-content)
[](https://www.sigmaaldrich.com/GB/en)
Products
Cart0
GBEN
Products
[Login / Register](https://www.sigmaaldrich.com/oidc-sign-in)
[Order Lookup](https://www.sigmaaldrich.com/GB/en/order-lookup)
[Quick Order](https://www.sigmaaldrich.com/GB/en/quick-order)
Cart0
[Home](https://www.sigmaaldrich.com/GB/en)[Viable Air Monitoring](https://www.sigmaaldrich.com/GB/en/applications/microbiological-testing/viable-air-monitoring)Settle Plates for Active and Passive Air Monitoring
# Settle Plates for Active and Passive Air Monitoring
Passive and active air sampling are essential methods for monitoring airborne microorganisms in controlled environments, particularly in the pharmaceutical and cosmetics industries.
__Passive air monitoring__ is usually performed by exposing the settle plates to the air for a given time and then incubating them to allow visible colonies to develop and be counted. The viable biological particles settle onto the plate's surface, allowing for qualitative analysis of airborne microorganisms once the plates are incubated.
__Active air monitoring__ with settle plates or contact plates utilizes a microbial air sampler to draw a predetermined volume of air over a collection medium, such as agar plates. The plate is then removed from the air sampler and directly incubated, allowing visible colonies to develop and subsequently be counted. The number of visible colonies provides an estimate of the number of colony-forming units in the sampled air.
__Section Overview__
- [ICR and ICRplus Gamma-Irradiated Settle Plates](https://www.sigmaaldrich.com#icr)
- [Non-Lockable ICR Settle Plates for Isolators and Critical Cleanrooms](https://www.sigmaaldrich.com#non-lockable)
- [Lockable ICRPlus Settle Plates for Isolators and Critical Cleanrooms](https://www.sigmaaldrich.com#lockable)
- [Non-Irradiated Settle Plates (LI Settle Plates) for Less Critical Areas](https://www.sigmaaldrich.com#non)
- [Explore Culture Media Products for Active and Passive Air Monitoring](https://www.sigmaaldrich.com#products)
We offer high-quality settle plates, including the gamma irradiated ICR and ICRplus plates, that are specifically designed for air monitoring in critical and less critical clean rooms. Our settle plates meet all relevant international standards and regulations, including EU cGMP, FDA Aseptic Guidance and USP 34<1116>. Our settle plates are easy-to-use, enabling continuous and efficient monitoring for microorganisms during production process.
[Contact Us](https://www.sigmaaldrich.com/GB/en/products/industrial-microbiology/viable-air-samplers/speak-to-a-specialist-microbial-air-sampling)
## [](https://www.sigmaaldrich.com)ICR AND ICRPLUS GAMMA-IRRADIATED SETTLE PLATES
ICR and ICRplus settle plates are designed to meet the demands of environmental monitoring in isolators and cleanrooms. The plates are gamma-irradiated and triple bagged to minimize the risk of cross-contamination, making them ideal for active air monitoring with our [](https://www.sigmaaldrich.com)[MAS Air Sampler](https://www.sigmaaldrich.com/GB/en/products/industrial-microbiology/viable-air-samplers#air)s and for passive microbial air sampling in isolators and clean rooms.
Our settle plates contain either Tryptic Soy Agar (TSA), Sabouraud Dextrose Agar (SDA) or a non-animal, vegetable peptone medium to reduce the risk of BSE/TSE contamination, all prepared in accordance with the recommendations of the European and US pharmacopeias. We offer Sabouraud Dextrose Agar formulations in pink plates to facilitate clear and easy differentiation of TSA and SDA.
__Figure 1.__Tripled bagged and gamma irradiated ICR and ICRplus settle plates
The plates are supplemented with neutralizers to inactivate a broad range of disinfectants and β-lactam antibiotics. To minimize the risk of contamination during transportation of the samples from a sterile to a non-classified environment, we provide gamma-sterilized zip lock bags suitable for contact and settle plates.
Additionally, the plates are available in both non-lockable ICR and lockable ICRplus configurations.
## [](https://www.sigmaaldrich.com)Non-lockable ICR settle plates for isolators and critical cleanrooms
Secure and reliableConvenientFlexible
- 2-D data matrix barcode on each plate for paperless and secure identification of individual plates
- H2O2-impermeable triple-sleeve packaging (transparent) for safe transfer into cleanrooms (ISO 5) and isolators. Protects media during decontamination of isolator
- Produced in cleanrooms, gamma-irradiated in final packaging to minimize the risk of contamination
- Storage at room temperature: storable at the site of use
- Long shelf life: Allows fewer incoming goods controls, cost-saving
- High filling volume settle plates (30 mL): Allows prolonged exposition and incubation
- SDA in pink plates easily distinguishes between TSA and SDA; Lessen the chance of mix up
- Usable for personnel monitoring or active air monitoring with air samplers
- Supplemented by neutralizers for a wide range of disinfectants and β-lactam antibiotics
- New media formulation of non-animal origin available: Minimizes the risk of BSE contamination.
## [](https://www.sigmaaldrich.com)Lockable ICRPlus settle plates for isolators and critical cleanrooms
__Figure 2.__Lockable ICRPlus settle plates
In addition to all the above features and benefits, our ICRPlus settle plates combine the high-quality features of standard ICR Settle Plates with the new two-way closure system (CLOSED- or VENT-position) to:
- Increase safety during transport between the sampling area and laboratory/incubation
- Expand incubation for the detection of anaerobic and microaerophilic microorganisms
## [](https://www.sigmaaldrich.com)NON-IRRADIATED SETTLE PLATES (LI SETTLE PLATES) for Less Critical Areas
Our range of non-irradiated settle plates is designed for use in non-controlled and less critically controlled cleanrooms. They can be used for passive and active air monitoring, as well as glove prints. Each set contains ten plates, which are single-bagged.
Single-bagged long (L) incubation (I) settle plates for environmental monitoring in less critical areas are ideal for monitoring less critical cleanroom areas, such as grade C, D, and E, or non-specified environments. Their high-filling volume of 30 mL in 90 mm settle plates reduces the percentage of water loss during the air monitoring procedures.
__Figure 3.__TSA and pink coloured SDA ICR plates
LI Settle Plates feature a higher than standard filling volume of 30 mL to compensate for water loss during extended incubation periods. These Settle Plates contain either Tryptic Soy Agar (TSA) or Sabouraud Dextrose Agar (SDA), which is ideally suited for the growth of yeast and molds are prepared according to the recommendations of the European and US pharmacopeias. They are supplemented with neutralizers such as Lecithin (L), Tween 80 (T), histidine (H) and sodium thiosulfate (Th) to inactivate a wide range of disinfectants. Additionally, to suppress bacterial growth, SDA media is made available with chloramphenicol.
Secure and reliableConvenientFlexible
- 2-D data matrix barcode on each plate for paperless and secure identification of individual plates
- Produced in cleanrooms to minimize the risk of contamination
- Prepared according to the recommendations of the European and US pharmacopeias
- Storage at room temperature: Storable at the site of use
- Long shelf life (6-9 months): Fewer incoming goods controls
- High-filling volume settle plates allow prolonged exposition and incubation
- Use for active and passive air monitoring, personnel monitoring
## [](https://www.sigmaaldrich.com)Explore Culture Media Products for Active and Passive Air Monitoring
## Tryptic Soy Agar
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
## Sabouraud Dextrose Agar
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
## CHOCOLATE AGAR +LTH - SDV
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
## Media and Accessories
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
* * *
__Related Articles__
- [Contact Plates and Swabs for Personnel and Surface Monitoring](https://www.sigmaaldrich.com/GB/en/technical-documents/technical-article/microbiological-testing/surface-and-personnel-testing/contact-plates-and-swabs)
- [IsoBag® Rapid Transfer Solution Optimizes Your Environmental Monitoring Workflow](https://www.sigmaaldrich.com/GB/en/technical-documents/technical-article/microbiological-testing/viable-air-monitoring/environmental-monitoring-isobags)
- [Aseptic Process Simulation in the Pharmaceutical Industry](https://www.sigmaaldrich.com/GB/en/technical-documents/technical-article/microbiological-testing/media-fills/media-fill-testing)
- [Media Fills Test for Beverages](https://www.sigmaaldrich.com/GB/en/technical-documents/technical-article/microbiological-testing/media-fills/linden-grain)
- [Bacterial Contamination Detection with ICR Swabs](https://www.sigmaaldrich.com/GB/en/technical-documents/technical-article/microbiological-testing/surface-and-personnel-testing/detection-of-low-bacterial-contamination)
- [Environmental Sample Pre Storage on Counting Results](https://www.sigmaaldrich.com/GB/en/technical-documents/technical-article/microbiological-testing/viable-air-monitoring/environmental-sample-pre-storage-on-counting-results)
__Related Product Categories__
- [Environmental Monitoring and Aseptic Process Simulation](https://www.sigmaaldrich.com/GB/en/products/industrial-microbiology/viable-air-samplers)
- [Microbial Culture Media](https://www.sigmaaldrich.com/GB/en/products/industrial-microbiology/microbial-culture-media)
Top
__Sign In To Continue__
Sign In__Don't Have An Account?__Register
An unknown error has occured.
- English - EN
[Learn More](https://www.sigmaaldrich.com)