Product Name
Acetylcholine chloride ≥99% (TLC)
Product Number
A6625
Product Brand
SIGMA
CAS Number
60-31-1
Molecular Weight
181.66
Test | Specification |
|---|---|
| APPEARANCE (COLOR) | WHITE TO WHITE WITH A FAINT YELLOW CAST |
| APPEARANCE (FORM) | Powder to Crystals |
| PURITY (TLC AREA %) | ≥ 99 % |
| SOLUBILITY (COLOR) | Colorless |
| SOLUBILITY (TURBIDITY) | Clear |
| SOLUBILITY (METHOD) | 400 MG IN 4 ML H2O |
| INFRARED SPECTRUM | CONFORMS TO STRUCTURE |