Merck
  • Efficient reduction of polycyclic quinones, hydroquinones, and phenols with hydriodic acid.
Efficient reduction of polycyclic quinones, hydroquinones, and phenols with hydriodic acid.
The Journal of Organic Chemistry (1979)
Konieczny M and Harvey RG.