ASTM D5134: GC Analysis of Hydrocarbons on Petrocol® DH 50.2
This application demonstrates the suitability of the Petrocol column for the efficient analysis of hydrocarbons as listed in ASTM method D5134.
ASTM D5134: GC Analysis of Hydrocarbons on Petrocol® DH 50.2
Conditions
Column Petrocol DH 50.2, 50 m x 0.20 mm I.D., 0.50 µm (24133-U)
Oven 35 °C (30 min), 2 °C/min to 200 °C (15 min)
Carrier Gas Helium, 19-21 cm/sec @ 35C (see ASTM D5134)
Injection 0.2 µL, 200:1 split
Injection Temperatur 200 °C
Detector FID, 250 °C
Description
Categories Analytical Chromatography, GC Applications, Hydrocarbons
Featured Industry Petroleum
Legal Information Petrocol is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
Suitability Application for GC
Analyte
  • 1,1-Dimethylcyclopentane
  • 1,1-Dimethylheptane
  • 1,4-Dimethylheptane
  • 1,5-Dimethylheptane
  • 2,2,3-Trimethylbutane
  • 2,2,3-Trimethylpentane
  • 2,2,5-Trimethylhexane
  • 2,2-Dimethylbutane
  • 2,2-Dimethylheptane
  • 2,2-Dimethylhexane
  • 2,2-Dimethylpentane
  • 2,3,5-Trimethylhexane
  • 2,3-Dimethylbutane
  • 2,3-Dimethylheptane
  • 2,3-Dimethylhexane
  • 2,3-Dimethylpentane
  • 2,4-Dimethylheptane
  • 2,4-Dimethylhexane
  • 2,4-Dimethylpentane
  • 2,5-Dimethylheptane
  • 2,5-Dimethylhexane
  • 2,6-Dimethylheptane
  • 2-Methyl-1-ethylpentane
  • 2-Methyl-4-ethylhexane
  • 2-Methylheptane
  • 2-Methylhexane
  • 2-Methyloctane
  • 2-Methylpentane
  • 3,3-Dimethylhexane
  • 3,3-Dimethylpentane
  • 3,4-Dimethylheptane
  • 3-Ethylheptane
  • 3-Ethylhexane
  • 3-Methyl-3-ethylpentane
  • 3-Methylheptane
  • 3-Methylheptane
  • 3-Methylhexane
  • 3-Methyloctane
  • 3-Methylpentane
  • 4-Ethylheptane
  • 4-Methylheptane
  • 4-Methyloctane
  • Benzene
  • cis-1,2-Ethylmethylcyclopentane
  • cis-1,3-Dimethylcyclopentane
  • cis-1,3-Ethylmethylcyclopentane
  • Cyclohexane
  • Cyclopentane
  • Ethylbenzene
  • Ethylcyclopentane
  • Iso-Butane
  • Isopentane
  • Methylcyclopentane
  • m-Xylene
  • n-Butane
  • n-Heptane
  • n-Hexame
  • n-Nonane
  • n-Octane
  • n-Pentane
  • o-Xylene
  • Propane
  • p-Xylene
  • Toluene
  • trans-1,1-Ethylmethylcyclopentane
  • trans-1,2-Dimethylcyclopentane
  • trans-1,2-Ethylmethylcyclopentane
  • trans-1,3-Dimethylcyclopentane