Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
ASTM D6526: GC Analysis of Impurities in Toluene on SLB®-IL100, 30 m Column
ASTM D6526: GC Analysis of Impurities in Toluene on SLB®-IL100, 30 m Column
Conditions
column SLB-IL100, 30 m x 0.25 mm I.D., 0.20 µm (28884-U)
oven 70 °C
carrier gas helium, 28 cm/sec
sample toluene, with each aromatic impurity spiked at 0.01% by weight
injection 1 µL40:1 split
liner 4 mm I.D., split type, cup design
inj. temp. 200 °C
detector FID, 200 °C
Description
Categories Analytical Chromatography, GC Applications, Aromatics, Hydrocarbons
Featured Industry Chemicals and Industrial Polymers
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC
suitability application for GC
Analyte
  • 1,4-Dioxane
  • Aliphatic hydrocarbons
  • Benzene
  • Ethylbenzene
  • Isopropylbenzene (cumene)
  • m-Xylene
  • o-Xylene
  • p-Xylene
  • Toluene