Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
GC Analysis for Aroma Fusel Alcohols in Vodka on Watercol™ 1910
GC Analysis for Aroma Fusel Alcohols in Vodka on Watercol™ 1910
Conditions
column Watercol 1910, 30 m x 0.25 mm I.D., 0.20 µm; two identical columns installed in the same inlet, each going to a separate detector (29711-U)
oven 50 °C (1 min), 5 °C/min to 180 °C (3 min)
carrier gas helium, 25 cm/sec (measured in each column at an oven temperature of 125 °C)
sample neat vodka
injection 1 µL, 100:1 split
liner 4 mm I.D., split/splitless type, wool packed single taper FocusLiner™ design
inj. temp. 250 °C
detector FID, 250 °C; TCD, 200 °C
Description
Analysis Note Fusel alcohols are by-products of the fermentation process. Four of these (n-propanol, isobutanol, active amyl alcohol, and isoamyl alcohol) are important to alcoholic beverage manufacturers due to their aroma characteristics.
The "Watercol" series of ionic liquid capillary GC columns are characterized by their ability to produce a sharp peak shape for water, sharp enough so that water can be integrated and quantified. Sharp peak shapes are also produced for many other small polar analytes, such as the four aroma fusel alcohols.
In this application, a vodka sample was analyzed for the presence of aroma fusel alcohols. As shown, none were detected.
Featured Industry Food and Beverages
Legal Information Watercol is a trademark of Sigma-Aldrich Co. LLC