Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
GC Analysis of 1-(α-Naphthyl)ethylamine Enantiomers (N-Trifluoroacetyl Derivatives) on Astec® CHIRALDEX® B-DA
GC Analysis of 1-(α-Naphthyl)ethylamine Enantiomers (N-Trifluoroacetyl Derivatives) on Astec® CHIRALDEX® B-DA
Conditions
column Astec CHIRALDEX B-DA, 30 m x 0.25 mm I.D., 0.12 µm (72023AST)
oven 160 °C
carrier gas helium, 35 psi
sample peak 1: R(+)-1-(α-naphthyl)ethylaminepeak 2: S(-)-1-(α-naphthyl)ethylamine
detector FID, 250 °C
Description
Categories Analytical Chromatography, GC Applications, Enantiomers, Anilines, Amines
Featured Industry Chemicals and Industrial Polymers
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC, CHIRALDEX is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes