Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
GC Analysis of 1-Phenylethylamine Enantiomers (N-Trifluoroacetyl Derivatives) on Astec® CHIRALDEX® B-DM
GC Analysis of 1-Phenylethylamine Enantiomers (N-Trifluoroacetyl Derivatives) on Astec® CHIRALDEX® B-DM
Conditions
column Astec CHIRALDEX B-DM, 30 m x 0.25 mm I.D., 0.12 µm (77023AST)
oven 120 °C
carrier gas helium, 30 psi
sample 3 mg/mL in methylene chloridepeak 1: R(+)-1-phenylethylamine (N-TFA derivative)peak 2: S(-)-1-phenylethylamine (N-TFA derivative)
injection 1 µL, 80:1 split
inj. temp. 250 °C
detector FID, 250 °C
Description
Analysis Note Can reverse elution order by using N-acetyl derivative on CHIRALDEX B-DM and B-PM, the N-chloroacetyl derivative on B-PM, or the N-TFA derivative on B-PH.
Categories Analytical Chromatography, GC Applications, Enantiomers, Anilines, Amines
Featured Industry Chemicals and Industrial Polymers
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC, CHIRALDEX is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes