Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
GC Analysis of 2-Butanol Enantiomers (O-Acetyl Derivatives) on Astec® CHIRALDEX® G-TA
GC Analysis of 2-Butanol Enantiomers (O-Acetyl Derivatives) on Astec® CHIRALDEX® G-TA
Conditions
column Astec CHIRALDEX G-TA, 30 m x 0.25 mm I.D., 0.12 µm (73033AST)
carrier gas helium, 30 psi
gradient 30 °C (5 min), 5 °C/min to 110 °C
sample 2 mg/mL each racemate in methylene chloridepeak 1: impuritypeak 2: (S)-2-butanol (O-acetyl derivative)peak 3: ®-2-butanol (O-acetyl derivative)
injection 1 µL, 80:1 split
inj. temp. 250 °C
detector FID, 250 °C
Description
Analysis Note Elution order reversal by using the O-TFA derivative on CHIRALDEX G-TA.
Categories Analytical Chromatography, GC Applications, Alcohols, Enantiomers
Featured Industry Chemicals and Industrial Polymers
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC, CHIRALDEX is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes