Attention:
As we update our core systems, key features of the website including ordering and real-time price and availability, may be unavailable from 8:00 PM CDT Friday, June 21st until 9:00 AM CDT Saturday morning June 22nd.
We apologize for any inconvenience this may cause.
GC Analysis of 2-Hydroxyoctanoic Acid Methyl Ester Enantiomers on Astec® CHIRALDEX® B-DP
GC Analysis of 2-Hydroxyoctanoic Acid Methyl Ester Enantiomers on Astec® CHIRALDEX® B-DP
Conditions
column Astec CHIRALDEX B-DP, 30 m x 0.25 mm I.D., 0.12 µm (78023AST)
oven 110 °C
carrier gas helium, 30 psi
sample peaks 1 & 2: 2-hydroxyoctanoic acid methyl ester enantiomers
inj. temp. 250 °C
detector FID, 250 °C
Description
Categories Analytical Chromatography, Chiral Applications, GC Applications, Clinical, Forensic & Toxicology, Acids, Biomarkers, Enantiomers
Featured Industry Chemicals and Industrial Polymers
Clinical
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC, CHIRALDEX is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes