Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
GC Analysis of a 12-Component Sulfur Compound Mix on SLB®-IL59
GC Analysis of a 12-Component Sulfur Compound Mix on SLB®-IL59
Conditions
column SLB-IL59, 30 m x 0.25 mm I.D., 0.20 µm (28891-U)
oven 30 °C (4 min), 10 °C/min to 300 °C
carrier gas helium, 30 cm/sec constant
sample 12 sulfur compounds, neat mix
injection 0.2 µLsplit 100:1
liner 4 mm I.D., split/splitless type, dual taper design
inj. temp. 250 °C
detector MSD, interface at 275 °C
Description
Categories Analytical Chromatography, GC Applications, Volatiles, Sulfur Compounds, Gases
Featured Industry Chemicals and Industrial Polymers
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC
suitability application for GC
Analyte
  • 2-Methylthiophene
  • Carbon disulfide
  • Diethyl disulfide
  • Diethyl sulfide
  • Dimethyl disulfide
  • Dimethylsulfide
  • Ethyl mercaptan
  • Ethyl methyl sulfide
  • Isopropyl mercaptan
  • n-Propyl mercaptan
  • Thiophane
  • Thiophene