GC Analysis of a 37-Component FAME Mix on SP™-2560
GC Analysis of a 37-Component FAME Mix on SP™-2560
column SP-2560, 100 m x 0.25 mm I.D., 0.20 µm (24056)
oven 140 °C (5 min), 8 °C/min to 180 °C, 4 °C/min to 210 °C, 20 °C/min to 250 °C (7 min)
carrier gas hydrogen, 40 cm/sec
sample Supelco 37-Component FAME Mix (47885-U), analytes at concentrations indicated in methylene chloride
injection 1 µL, 100:1 split
liner 4 mm I.D., cup design
inj. temp. 250 °C
detector FID, 250 °C
Categories Analytical Chromatography, GC Applications, Fatty Acid Methyl Esters (FAMEs)
Featured Industry Agriculture
Food and Beverages
Legal Information SP is a trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Gas Purification and Gas Management