Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
GC Analysis of Alcohol Enantiomers on β-DEX™ 325
GC Analysis of Alcohol Enantiomers on β-DEX™ 325
Conditions
column beta-DEX 325, 30 m x 0.25 mm I.D., 0.25 µm (24308)
oven 110 °C, 1-phenyl-2-propanol; 100 °C, isopinocampheol; 90 °C, 6-methyl-5-hepten-2-ol
carrier gas helium, 20 cm/sec @ analysis temperature
sample 1 mg/mL in methylene chloride
injection 1 µL100:1 split
inj. temp. 220 °C
detector FID, 300 °C
Description
Categories Analytical Chromatography, GC Applications, Enantiomers, Alcohols
Featured Industry Chemicals and Industrial Polymers
Legal Information DEX is a trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • 1-Phenyl-2-propanol
  • 6-Methyl-5-hepten-2-ol
  • Isopinocampheol