Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
GC Analysis of Alkylamine Enantiomers (N-Trifluoroacetyl Derivatives) on Astec® CHIRALDEX® B-TA
GC Analysis of Alkylamine Enantiomers (N-Trifluoroacetyl Derivatives) on Astec® CHIRALDEX® B-TA
Conditions
column Astec CHIRALDEX B-TA, 30 m x 0.25 mm I.D., 0.12 µm (73023AST)
carrier gas helium, 30 psi
gradient 80 °C, 2 °C/min to 110 °C
sample 2.5 mg/mL each racemate in methanol (N-TFA derivatives)
injection 0.6 µL, 80:1 split
inj. temp. 250 °C
detector FID, 250 °C
Description
Categories Analytical Chromatography, GC Applications, Enantiomers, Amines
Featured Industry Chemicals and Industrial Polymers
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC, CHIRALDEX is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes