GC Analysis of Anilines on Equity®-5
GC Analysis of Anilines on Equity®-5
column Equity-5, 30 m x 0.25 mm I.D., 0.25 µm (28089-U)
oven 50 °C (2 min), 10 ºC/min to 200 °C, 15 ºC/min to 325 °C
carrier gas helium, constant flow, 1.3 mL/sec
sample 50 ng on-column of a custom anilines mix
injection 1 µL, splitless (0.5 min)
liner 4 mm I.D., single taper
inj. temp. 250 °C
detector FID, 325 °C
Categories Analytical Chromatography, GC Applications, Anilines
Featured Industry Chemicals and Industrial Polymers
Legal Information Equity is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
  • 2,4,5-Trichloroaniline
  • 2,4-Diaminotoluene
  • 2,6-Diethylaniline
  • 2,6-Dimethylaniline
  • 2-Chloroaniline
  • 2-Methyl-6-ethylaniline
  • 3,3′-Dichlorobenzidine
  • 3,4-Dichloroaniline
  • 3-Aminobenzotrifluoride
  • 3-Chloro-4-fluoroaniline
  • 4-Chloro-2-nitroaniline
  • 4-Chloro-4-methylaniline
  • 4-Isopropylaniline
  • Aniline
  • N,N-Dimethylaniline
  • o-Toluidine
Related Links