Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
GC Analysis of Aromatics in Reformulated Gasoline (Ethanol at 20%) on SLB®-IL111
GC Analysis of Aromatics in Reforµlated Gasoline (Ethanol at 20%) on SLB®-IL111
Conditions
column SLB-IL111, 60 m x 0.25 mm I.D., 0.20 µm (28928-U)
oven 50 °C (3 min), 15 °C/min to 265 °C (5 min)
carrier gas helium, 30 cm/sec
sample Premium unleaded gasoline, plus ethanol at 20% (v/v) and two internals each at 4% (v/v)
injection 0.5 µL, 200:1 split
liner 4 mm I.D., split type, single taper wool packed FocusLiner™ design
inj. temp. 250 °C
detector FID, 275 °C
Description
Categories Analytical Chromatography, GC Applications, Aromatics, Hydrocarbons
Featured Industry Petroleum
Legal Information FocusLiner is a trademark of SGE Analytical Science Pty Ltd., SLB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes