GC Analysis of Aromatics in Reformulated Gasoline on TCEP
GC Analysis of Aromatics in Reforµlated Gasoline on TCEP
column TCEP, 60 m x 0.25 mm I.D., 0.44 µm (24153)
oven 50 °C (3 min), 10 °C/min to 140 °C (35 min)
carrier gas helium, 30 cm/sec constant
sample Reforµlated gasoline
injection 1 µL, 100:1 split
liner 4 mm I.D. FocusLiner™ inlet liner with taper
inj. temp. 220 °C
detector FID, 170 °C
Categories Analytical Chromatography, GC Applications, Aromatics, Hydrocarbons
Featured Industry Petroleum
Legal Information FocusLiner is a trademark of SGE Analytical Science Pty Ltd.
Other Notes Complementary products:
Gas Purification and Gas Management