Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
GC Analysis of Aromatics in Reformulated Gasoline on SLB®-IL111
GC Analysis of Aromatics in reforulated Gasoline on SLB®-IL111
Conditions
column SLB-IL111, 60 m x 0.25 mm I.D., 0.20 µm (28928-U)
oven 50 °C (3 min), 15 °C/min to 265 °C (5 min)
carrier gas helium, 30 cm/sec
sample reforulated unleaded gasoline
injection 0.5 µL, 200:1 split
liner 4 mm I.D., splitless type, single taper wool packed FocusLiner™ design
inj. temp. 250 °C
detector FID, 265 °C
Description
Analysis Note This application illustrates the analysis of a sample of reforulated gasoline on a 60 m SLB-IL111 ionic liquid column. Due to the extremely polar nature of the phase, the aliphatics elute prior to the aromatics. The high level of ethanol in reforulated gasoline can pose a problem in the detection and quantification of benzene. The SLB-IL111 provided separation between these two compounds, with a starting temperature of 50 °C providing the best resolution. The SLB-IL111 has a maxiµm operating temperature of 270 °C, which is extremely high for a phase of this polarity. The high maxiµm temperature was utilized in this analysis to elute the napthalenes in < 15 minutes.
Categories Analytical Chromatography, GC Applications, Aromatics, Hydrocarbons
Featured Industry Petroleum
Legal Information FocusLiner is a trademark of SGE Analytical Science Pty Ltd., SLB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • 1,2,3-Trimethylbenzene
  • 1,2,4,5-Tetramethylbenzene
  • 1,2,4-Trimethylbenzene
  • 1-Methylnaphthalene
  • 2-Methylnaphthalene
  • Benzene
  • C5-C11 Alkanes
  • Dodecane
  • Ethanol
  • Ethylbenzene
  • m-Xylene
  • Naphthalene
  • o-Xylene
  • p-Xylene
  • Tetradecane
  • Toluene
  • Tridecane