Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
GC Analysis of Bacterial Acid Methyl Esters (BAMEs) on SPB®-1
GC Analysis of Bacterial Acid Methyl Esters (BAMEs) on SPB®-1
Conditions
column SPB-1, 30 m x 0.25 mm I.D., 0.25 µm film (24028)
oven 150 C (4 min) to 250 C (5 min) at 4 C
carrier gas helium, 20 cm/sec (150 C)
inj. temp. 1 µL
detector FID
Description
Categories Analytical Chromatography, GC Applications, Bacterial Acid Methyl Esters (BAMEs)
Featured Industry Clinical
Legal Information SPB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • Methyl 12-methyltetradecanoate
  • Methyl 13-methyltetradecanoate
  • Methyl 14-methylpentadecanoate
  • Methyl 15-methylhexadecanoate
  • Methyl 2-hydroxydodecanoate
  • Methyl 2-hydroxydodecanoate
  • Methyl 2-hydroxyhexadecanoate
  • Methyl 2-hydroxytetradecanoate
  • Methyl 3-hydroxydodecanoate
  • Methyl 3-hydroxytetradecanoate
  • Methyl cis-9-hexadecenoate
  • Methyl dodecanoate
  • Methyl heptadecanoate
  • Methyl hexadecenoate
  • Methyl nonadecanoate
  • Methyl pentadecanoate
  • Methyl tetradecanoate
  • Methyl tridecanoate
  • Methyl undecanoate
  • Methyl cis-9,10-methylenehexadecanoate
  • Methyl cis-9,10-methyleneoctadecanoate
  • Methyl cis-9,12-octadecadienoate
  • Methyl cis-9-octadecenoate
  • Methyl eicosanoate
  • Methyl octadecenoate
  • Methyl trans-9-octadecenoate